Deionizer, B-Pure™
Supplier: Thermo Fisher Scientific
D4511 D4505
BARND4511EA 576 EUR
BARND4511 BARND4505
Deionizer, B-Pure™
Water Purification Systems Deionisers
These B-Pure™ laboratory-grade water systems are designed for pre-treatment or deionization application needs. The design provides the ability to add additional holders when the need arises. Choose from single, double or half-size holders.
  • Easily change cartridges with the simple quarter turn quick release canisters
  • Quarter-turn quick-release canisters make cartridge changes easy
  • Accepts Pura-Lite™ indicator or digital purity meter to monitor water quality
Single module: Simple system without draw-off valve or purity indicator and produces up to 4 l/More Product Information
Order Now
SPECIFICATIONS
Model D4511 D4505
Environmentally Preferable
Max. flow rate 240 l/h 120 l/h
W×D×H 178×178×610 mm 178×178×381 mm