Digital hotplate, SuperNuova+™
Supplier: Thermo Fisher Scientific
HP88857195 HP88857198 HP88850195
BARNHP88857195EA 920 EUR
BARNHP88857195 BARNHP88857198 BARNHP88850195
Digital hotplate, SuperNuova+™
Heaters Hotplates
These hotplates features a raised display design which protects the electronics from any liquid spills and units are available with a choice of ceramic or aluminium top plates.
  • Easy-to-read LED display for heating
  • Hot Top warning system protects from accidental burns
  • Temperature adjustable in 1 degree increments
  • 12 litre capacity
  • Heating up time is 8 min to maximum temperature
Temperature control accuracy (at 37 °C): ≤±5 °C
Temperature uniformity (at 37 °C): ≤±2 °C
Delivery information: Supplied with cCSAUS certificate. Other accessories such asMore Product Information