CytoFLEX nano Flow Cytometers
Supplier: Beckman Coulter
D00710 D02028
BECMD00710EA 0 EUR
BECMD00710 BECMD02028
CytoFLEX nano Flow Cytometers
Cell Counters and Flow Cytometers Flow Cytometers
The CytoFLEX nano Flow Cytometer is the newest member of the CytoFLEX platform.
  • Specifically designed for the analysis of nanoparticles
  • CytoFLEX nano Flow Cytometer overcomes the limitations of traditional flow cytometry
  • Provides researchers a sensitive, consistent, and flexible solution to advance nanoparticle research
The CytoFLEX nano Flow Cytometer is a pivotal development in flow cytometry that allows users to analyse extracellular vesicles (EVs) at least as small as 40 nm with ease. It simultaneously offers up to