Buchner funnel trowel, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
368200118
BELA368200118EA 222 EUR
BELA368200118
Buchner funnel trowel, SP Bel-Art
Funnels Filtration Funnels
Specifically designed for use with 18, 24 and 36” Buchner Funnels, apply slight downward pressure and ‘sweep’ the trowel along the surface of the precipitate to quickly and efficiently smooth fissures appearing during the filtration process.
  • Large surface area smooths fissures faster than the traditional narrow spatula method
  • Beveled edges on both sides permit bi-directional smoothing
  • Curved ends match Buchner Funnel sidewalls allowing easy access to the entire cake surface
  • FDA-grade high-density polyethylene (HDPE) complies with industry standards and