Racks for PCR and microcentrifuge tubes, double-sided, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
F188450001 F188450002 F188450012
BELAF188450001EA 124 EUR
BELAF188450001 BELAF188450002 BELAF188450012
Racks for PCR and microcentrifuge tubes, double-sided, SP Bel-Art
Racks Tube Racks
PP, coloured.
  • Compact rack for benchtop use or freezer storage down to −80 °C
  • One side holds PCR tubes and the other 1,5 to 2,0 ml microtubes
  • Array is numbered for easy sample identification
  • Molded in handles provide a secure grip for transport and a matte side panel accepts tape or direct labelling
Assorted pack includes each one of yellow, red, blue, yellow, pink.