Petri dish rack, 60 mm, Bel-Art ProCulture, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
F189910060
BELAF189910060EA 218 EUR
BELAF189910060
Petri dish rack, 60 mm, Bel-Art ProCulture, SP Bel-Art
Racks Petri Dish Racks
This rack is useful for safe transport and incubation of 60 mm Petri dishes and helps to streamline benchtop sample processing. Rack base is epoxy coated steel, and all other parts are made of white polypropylene.
  • Six columns hold up to fifty-four Petri dishes with lids
  • 10½×6³/₄" base area preserves benchtop space
  • Large center divider has space for labeling; includes a carrying handle that does not interfere with samples
  • Up to four racks can be stacked together and interlocked to prevent accidental knock over
  • Finger access slots in the base allow a stack