Reagent digger, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
F367130000
BELAF367130000EA 58.5 EUR
BELAF367130000
Reagent digger, SP Bel-Art
Samplers Solid Samplers
Stainless steel. This robust tool scrapes and scoops caked samples from all vessels.
  • Ideal for retrieving flaked, powdered or granular media
  • V-shape directs sample dispensing
  • Fits into bottles with minimum neck diameter of 24 mm