Hand protectors, Hot Hand®, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
F380000000 F380000001 F380000002
112-1982EA 68.2 EUR
112-1982 BELAF380000001 BELAF380000002
Hand protectors, Hot Hand®, SP Bel-Art
Hot Hand Protectors
This protector is made of durable silicone rubber which remains flexible from –57 to +260 °C.
  • Safe, non-slip protection against very hot or cold surfaces
  • End pockets accept thumb and fingers to grasp objects
  • Multiple studs with concave tips cover surface of the Mitt, providing secure grip on virtually all hot/cold surfaces
  • One size fits all hands, left or right
Dimensions: 100×190 mm