Magnetic stirring bars positioner/retrievers, Spinbar®, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
H377800000
BELAH377800000EA 154 EUR
BELAH377800000
Magnetic stirring bars positioner/retrievers, Spinbar®, SP Bel-Art
Stirrers Stirring Tools
PE, to retrieve stirbars without coming into contact with the liquid in the containers.
  • No direct contact with stirring magnet
  • Splash-free operation