Slide tray and cabinet, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
H445830000 H445810000
BELAH445830000EA 1150 EUR
BELAH445830000 BELAH445810000
Slide tray and cabinet, SP Bel-Art
Slide Holders
This tray and cabinet system can hold up to 240 slides in a dust-free enclosure.
  • Cabinet and trays sold separately
  • White PS background to distinguish dyes
  • Handy finger cut out simplifies removal of slides
Informace pro objednávání: Includes PP outer shell, PS tray rails and a vertically sliding, clear acrylic door with a knob for easy opening.