Cleanroom wipes, Twillx® 1622
Supplier: BERKSHIRE
T1622.0909.8 T162212124/150
BERKT1622.0909.8EA 3070 EUR
BERKT1622.0909.8 BERKT162212124/150
Cleanroom wipes, Twillx® 1622
Wipes Cleanroom Wipes Dry Cleanroom Wipes
These environmentally friendly wipes made of 100% woven cotton are engineered for thermal stability and high heat applications.
  • Highly sorbent and durable
  • Work well with IPA and other solvents
  • Bias cut and tight weave construction
  • Excellent resistance to the buildup of electrostatic discharge in 40-60% RH environments
  • Autoclavable
Ideal for cleaning high temperature equipment and diffusion furnaces, for polishing and burnishing metallic and non metallic magnetic media disk surfaces and for cleaning solvent spills.
Certifikace: ISO Class 6 to 8,More Product Information