Anti-BHLHB5 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB155838-200
BIRBORB155838-200EA 0 EUR
BIRBORB155838-200
Anti-BHLHB5 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: BHLHB5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR