Filter tips, uTIP™
Supplier: BIOTIX
M-1250-9FC96 M-0200-9FC 63300154 63300041 63300042 63300043 63300045 63300047 63300153 63300044 63300046
732-2383EA 220 EUR
732-2383 732-3397 732-1429 BITX63300041 BITX63300042 BITX63300043 BITX63300045 BITX63300047 BITX63300153 BITX63300044 BITX63300046
Filter tips, uTIP™
Pipette Tips
This new generation of filter tips form an excellent seal on both single- and multi-channel pipettes. The tips are manufactured with proprietary technologies for increased pipetting accuracy and precision. Biotix filters are made from only the best materials and keep pipettes free of aerosol contamination.
  • PP, sterile, transparent
  • FlexFit™ provides a flexible nature at the proximal end of the tip, so reducing the loading and ejection forces when using a pipette
  • Blade™ technology eliminates hanging drops at the distal end of the pipette tip
  • Certified free