L-Methionine

Supplier: BIOMOL RESEARCH LABORATORIES

Synonyms: (S)-(-)-2-Amino-4-(methylthio)butyric acid, (S)-(-)-Methionine, L(-)-Methionine, (S)-(-)-2-Amino-4-(methylthio)butanoic acid, H-Met-OH, Methionine, (S)-2-Amino-4-(methylthio)butanoic acid

06023-1 06023-2
BMOL06023-1EA 0 EUR
BMOL06023-1 BMOL06023-2
L-Methionine
L-Methionine
Formula: C₅H₁₁NO₂S
MW: 149.21 g/mol
Boiling Pt: 278
Melting Pt: 136
Density: 1.373 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00063097
CAS Number: 63-68-3
EINECS: 200-562-9
Merck Index: 13,06004

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR