Sodium acetate

Supplier: BIOMOL RESEARCH LABORATORIES

Synonyms: Acetic acid sodium salt

54652-2 54652-3
BMOL54652-2EA 0 EUR
BMOL54652-2 BMOL54652-3
Sodium acetate
Sodium acetate
Formula: H₃CCOONa
MW: 82.03 g/mol
Boiling Pt: 881.4
Melting Pt: 324
Density: 1.528 g/cm³ (20 °C)
Flash Pt: >250 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00012459
CAS Number: 127-09-3
EINECS: 204-823-8
REACH: 01-2119485123-42
Merck Index: 12,08711

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR