Stirrer bearings, PTFE
Supplier: Bohlender
C424-04 C424-05 C424-08 C424-09 C424-12 C424-13 C424-14 C424-16 C424-18 C429-18 C429-14
BOHLC424-04EA 42.9 EUR
BOHLC424-04 BOHLC424-05 BOHLC424-08 BOHLC424-09 BOHLC424-12 BOHLC424-13 BOHLC424-14 BOHLC424-16 BOHLC424-18 BOHLC429-18 BOHLC429-14
Stirrer bearings, PTFE
Stirrers Stirring Tools
These bearings have sealing rings to ensure a perfect seal. A special gasket made of PTFE and a FPM o-ring which is compressed by a GL screw cap ensure the stirrer shaft is sealed.
  • Cone can be removed easily from the socket
  • Ground joint does not stick and danger of breaking is reduced
  • Gasket can be exchanged when worn