HPLC distributors for bottles
Supplier: Bohlender
D606-08
BOHLD606-08EA 19.2 EUR
BOHLD606-08
HPLC distributors for bottles
Closures Bottle Closures
Green screw cap, made of glass-fibre reinforced PP, and distributor body made of PP.
  • Very good chemical resistance
  • Temperature resistant from −50 to +100 °C
Suitable for GL 45 threaded bottles and it is available with the plugs for closing unused ports.