Double spatulas, BOLA
Supplier: Bohlender
H915-06 H915-02 H915-04
BOHLH915-06EA 18.9 EUR
BOHLH915-06 BOHLH915-02 BOHLH915-04
Double spatulas, BOLA
Spatulas
PTFE, white.
  • With tapered ends
  • Universal chemical resistance
  • Temperature resistant from −200 to +250 °C
  • Suitable for food contact (FDA conform)