Anti-APAF1 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss
BS-0059R-CY5
BOSSBS-0059R-CY5EA 900 EUR
BOSSBS-0059R-CY5
Anti-APAF1 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Oligomeric Apaf-1 mediates the cytochrome c-dependent autocatalytic activation of pro-caspase-9 (Apaf-3), leading to the activation of caspase-3 and apoptosis. This activation requires ATP. Isoform 6 is less effective in inducing apoptosis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: APAF1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

SPECIFICATIONS

Antigen symbol APAF1
Antigen name Apoptotic Peptidase Activating Factor 1
Antigen synonyms DKFZp781B1145|CED4|KIAA0413
Antibody type Primary
Clonality Polyclonal
Conjugation Cy5®
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 317
Isotype IgG
ImmunoChemistry Yes
Immunogen 3-44/1248
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR