Anti-CT Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Supplier: Bioss
BS-0063R-A680
BOSSBS-0063R-A680EA 890 EUR
BOSSBS-0063R-A680
Anti-CT Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
Calcitonin causes a rapid but short-lived drop in the level of calcium and phosphate in blood by promoting the incorporation of those ions in the bones. Katacalcin is a potent plasma calcium-lowering peptide.

Type: Primary
Antigen: Calcitonin
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 121-141/141
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR