Anti-CASP6 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss
BS-0084R-A350
BOSSBS-0084R-A350EA 893 EUR
BOSSBS-0084R-A350
Anti-CASP6 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Involved in the activation cascade of caspases responsible for apoptosis execution. Cleaves poly(ADP-ribose) polymerase in vitro, as well as lamins. Overexpression promotes programmed cell death.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CASP6
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR