Anti-GTP Cyclohydrolase 1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Supplier: Bioss
BS-0136R-A750
BOSSBS-0136R-A750EA 890 EUR
BOSSBS-0136R-A750
Anti-GTP Cyclohydrolase 1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antibodies
GTP cyclohydrolase I (also designated dopa-responsive dystonia) catalyses the conversion of GTP to D-erythro-7,8-dihydroneopterin triphosphate, the first and rate-limiting step in tetrahydrobiopterin (BH4) biosynthesis. Tetrahydrobiopterin is an essential cofactor for 3 aromatic amino acid monooxygenases: phenylalanine, tyrosine, and tryptophan hydroxylases. Animals can synthesise tetrahydrobiopterin in vivo from GTP through several enzymatic reactions.

Type: Primary
Antigen: GTP cyclohydrolase 1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 34-110/250
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR