Anti-RARA Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss
BS-0251R-A647
BOSSBS-0251R-A647EA 893 EUR
BOSSBS-0251R-A647
Anti-RARA Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
Receptor for retinoic acid. Retinoic acid receptors bind as heterodimers to their target response elements in response to their ligands, all-trans or 9-cis retinoic acid, and regulate gene expression in various biological processes. The RXR/RAR heterodimers bind to the retinoic acid response elements (RARE) composed of tandem 5'-AGGTCA-3' sites known as DR1-DR5. In the absence of ligand, the RXR-RAR heterodimers associate with a multiprotein complex containing transcription corepressors that induce histone acetylation, chromatin condensation and transcriptional suppression. On ligand binding,More Product Information
Order Now

SPECIFICATIONS

Antigen symbol RARA
Environmentally Preferable
Antigen name Retinoic Acid Receptor Alpha
Antigen synonyms Retinoic acid receptor|nucleophosmin-retinoic acid receptor alpha fusion protein NPM-RAR long form|NR1B1|alpha polypeptide|RAR|nucleophosmin-retinoic acid receptor alpha fusion protein NPM-RAR short form
Antibody type Primary
Clonality Polyclonal
Conjugation Alexa Fluor® 647
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 19401
Isotype IgG
ImmunoChemistry Yes
Immunogen 410-462/462
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR