Anti-ESE1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss
BS-11279R-HRP
BOSSBS-11279R-HRPEA 900 EUR
BOSSBS-11279R-HRP
Anti-ESE1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
ESE-1, a member of the Ets family of transcription factors, critically regulates epithelial cell differentiation and mediates vascular inflammation. ESE-1 is strongly expressed in vascular endothelium and smooth muscle cells where it is induced in response to inflammatory cytokines and lipopolysaccharides, interacts with NF-KappaB to induce nitric oxide synthase, and is induced during terminal differentiation of epidermal and primary keratinocytes. In addition, ESE-1 is upregulated upon differentiation of corneal epithelium and interacts with Sp1 and AP-1 proteins to induce squamousMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol ESE1
Antigen name ETS Domain Transcription Factor, Serine Box, Epithelial-Specific
Environmentally Preferable
Antigen synonyms Ets domain transcription factor serine box|ETS-related transcription factor Elf-3|Epithelium restricted Ets protein ESX|E74 like factor 3 ets domain transcription factor epithelial specic|Epithelium-restricted Ets protein ESX|E74-like factor 3|Ets transcription factor|EPR1|E74 like factor 3 ETS domain transcription factor serine box epithelial specic|E74 like factor 3 ets domain transcription factor|Epithelium specic Ets factor 1|MGC139318.|Epithelium specic Ets transcription factor 1|ESX|Epithelial-restricted with serine box|ERT|Elf3|ELF3_HUMAN|jen|ESE-1|EPR 1|Epithelial restricted with serine box|Epithelium-specic Ets transcription factor 1|E74 like factor 3|Ets domain transcription factor serine box epithelial specic
Antibody type Primary
Clonality Polyclonal
Conjugation HRP (Horseradish Peroxidase)
Reactivity Human,Rat,Mouse
Host Rabbit
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR