Anti-CSDC2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss
BS-11823R-A350
BOSSBS-11823R-A350EA 893 EUR
BOSSBS-11823R-A350
Anti-CSDC2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
PIPPIN is a Y-box protein (also called cold-shock (CSD) domain-containing protein) and belongs to a family of highly conserved RNA-binding transcriptional regulators. Predominantly expressed in brain cells and localizing to the nucleus and the cytoplasm, PIPPIN contains two RNA-binding motifs, namely PIP1 and PIP2, and one CSD domain. PIPPIN functions as a nucleic acid binding regulatory factor and is believed to participate in brain maturation. More specifically, PIPPIN binds to the 3’-UTR ends of the mRNAs encoding Histone H1 and Histone H3.3. This interaction requires all of the PIPPINMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol CSDC2
Antigen name Cold Shock Domain Containing C2
Environmentally Preferable
Antigen synonyms RNA binding protein PIPPin|Csdc2|Cold shock domain containing C2 RNA binding|CSDC2_HUMAN|Cold shock domain-containing protein C2|RNA-binding protein PIPPin.|Cold shock domain containing protein C2|dJ347H13.2|PIPPIN|CSDC 2
Antibody type Primary
Clonality Polyclonal
Conjugation Alexa Fluor® 350
Reactivity Human,Rat,Mouse
Host Rabbit
Isotype IgG
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR