Anti-GPRC5D Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss
BS-12169R-CY7
BOSSBS-12169R-CY7EA 900 EUR
BOSSBS-12169R-CY7
Anti-GPRC5D Rabbit Polyclonal Antibody (Cy7®)
Antibodies
The protein encoded by this gene is a member of the G protein-coupled receptor family; however, the specific function of this gene has not yet been determined. [provided by RefSeq, Jul 2008].

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: GPRC5D
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol GPRC5D
Environmentally Preferable
Antigen name G Protein-coupled Receptor Class C Group 5 Member D
Antigen synonyms GPC5D_HUMAN|G-protein coupled receptor family C group 5 member D|GPCR GPRC5D.|G protein coupled receptor family C group 5 member D|GPRC5D
Antibody type Primary
Clonality Polyclonal
Conjugation Cy7®
Reactivity Human
Host Rabbit
Isotype IgG
ImmunoChemistry Yes
Flow cytometry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR