Anti-CDON Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss
BS-12323R-CY3
BOSSBS-12323R-CY3EA 900 EUR
BOSSBS-12323R-CY3
Anti-CDON Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Cell adhesion molecule-related/down-regulated by oncogenes (CDO) and BOC (brother of CDO) are members of the immunoglobulin/fibronectin type III repeat family and act as cell surface receptors. CDO is a component of a cell-surface receptor complex which also contains BOC, NEO1, CTNNB1 and cadherins and which acts as a mediator of cell-cell interactions between muscle cells. CDO and BOC are single pass membrane proteins that play a role in myogenic cell differentiation. Together, CDO and BOC participate in a positive feedback loop with MyoD, a myogenic transcription factor. The 1,242 aminoMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol CDON
Antigen name Cell Adhesion Associated, Oncogene regulated
Environmentally Preferable
Antigen synonyms Cell adhesion molecule-related/down-regulated by oncogenes|CDON|CDO|CDON_HUMAN.
Antibody type Primary
Clonality Polyclonal
Conjugation Cy3®
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 50937
Isotype IgG
ImmunoChemistry Yes
Flow cytometry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR