Anti-CYP26A1 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss
BS-12928R-CY7
BOSSBS-12928R-CY7EA 900 EUR
BOSSBS-12928R-CY7
Anti-CYP26A1 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
The cytochrome P450 proteins (CYPs) are monooxygenases that catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids, and other lipids. P450 enzymes are classified into subfamilies based on their sequence similarities. CYP26A1 is a major retinoic acid catabolic enzyme. CYP26A1 plays an important role in protecting tailbud tissues from inappropriate exposure to retinoic acid. CYP26A1 transcription is epigenetically regulated by nuclear retinoic acid receptor ∫2. Mutations in the gene encoding for CYP26A1 are associated with caudal agenesis and spina bifida,More Product Information
Order Now

SPECIFICATIONS

Antigen symbol CYP26A1
Environmentally Preferable
Antigen name Cytochrome P450, Family 26, Subfamily A, Polypeptide 1
Antigen synonyms CP26|P450RAI|P450RAI1|CYP26
Antibody type Primary
Clonality Polyclonal
Conjugation Cy7®
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 1592
Isotype IgG
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR