Anti-CSP Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Supplier: Bioss
BS-12944R-A750
BOSSBS-12944R-A750EA 890 EUR
BOSSBS-12944R-A750
Anti-CSP Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antibodies
Cysteine string proteins (CSPs) are synaptic vesicle-associated, secretory vesicle proteins that are involved in Ca²⁺-regulated exocytosis of synaptic vesicles and modulation of presynaptic transmembrane calcium fluxes in neuroendocrine and endocrine cell types. CSP contains a J-domain that binds HSP 70/HSC 70 chaperone ATPases and a membrane-targeting, palmitoylated cysteine-rich string region. CSPs may act as molecular chaperones in synapses, and mediate conformational folding of components of the vesicular exocytotic machinery. CSP is involved in the fine tuning of neurotransmissionMore Product Information