Anti-DNAJC5 Rabbit Polyclonal Antibody (Cy7®)
Supplier: Bioss
BS-12944R-CY7
BOSSBS-12944R-CY7EA 900 EUR
BOSSBS-12944R-CY7
Anti-DNAJC5 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Cysteine string proteins (CSPs) are synaptic vesicle-associated, secretory vesicle proteins that are involved in Ca2+-regulated exocytosis of synaptic vesicles and modulation of presynaptic transmembrane calcium fluxes in neuroendocrine and endocrine cell types. CSP contains a J-domain that binds HSP 70/HSC 70 chaperone ATPases and a membrane-targeting, palmitoylated cysteine-rich string region. CSPs may act as molecular chaperones in synapses, and mediate conformational folding of components of the vesicular exocytotic machinery. CSP is involved in the fine tuning of neurotransmissionMore Product Information