Anti-DHRS7B Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss
BS-13000R-A488
BOSSBS-13000R-A488EA 1350 EUR
BOSSBS-13000R-A488
Anti-DHRS7B Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
This gene is located within the Smith-Magenis syndrome region on chromosome 17. It encodes a protein of unknown function. [provided by RefSeq, Jul 2008].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DHRS7B
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR