Anti-ITPA Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss
BS-13590R-A555
BOSSBS-13590R-A555EA 893 EUR
BOSSBS-13590R-A555
Anti-ITPA Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
ITPase (inosine triphosphate pyrophosphatase) is also known as putative oncogene protein hlc14-06-p or ITPA (inosine triphosphatase (nucleoside triphosphate pyrophosphatase)) and is a 194 amino acid protein. ITPase is abundantly expressed in heart, liver, sex glands, thyroid and adrenal gland, and is localized to the cytoplasm in the cell. ITPase catalyzes the pyrophosphohydrolysis of both ITP (inosine triphosphate) and dITP (deoxyinosine triphosphate) to IMP (inosine monophosphate) and diphosphate. IMP can be used as a substrate for purine nucleotide pathways. IMP can be phosphorylated toMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol ITPA
Antigen name Inosine Triphosphatase
Environmentally Preferable
Antigen synonyms My049|C20orf37|dJ794I6.3|HLC14-06-P
Antibody type Primary
Clonality Polyclonal
Conjugation Alexa Fluor® 555
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 3704
Isotype IgG
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR