Anti-COQ6 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Supplier: Bioss
BS-13987R-A750
BOSSBS-13987R-A750EA 1350 EUR
BOSSBS-13987R-A750
Anti-COQ6 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antibodies
FAD-dependent monooxygenase required for the C5-ring hydroxylation during ubiquinone biosynthesis. Catalyzes the hydroxylation of 3-polyprenyl-4-hydroxybenzoic acid to 3-polyprenyl-4,5-dihydroxybenzoic acid. The electrons required for the hydroxylation reaction may be funneled indirectly from NADPH via a ferredoxin/ferredoxin reductase system to COQ6.

Type: Primary
Antigen: COQ6
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 51-150/468
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR