Anti-TERT Rabbit Polyclonal Antibody (Cy5®)
Supplier: Bioss
BS-1411R-CY5
BOSSBS-1411R-CY5EA 883 EUR
BOSSBS-1411R-CY5
Anti-TERT Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Telomerase is a ribonucleoprotein enzyme essential for the replication of chromosome termini in most eukaryotes. Active in progenitor and cancer cells. Inactive, or very low activity, in normal somatic cells. Catalytic component of the teleromerase holoenzyme complex whose main activity is the elongation of telomeres by acting as a reverse transcriptase that adds simple sequence repeats to chromosome ends by copying a template sequence within the RNA component of the enzyme. catalyses the RNA-dependent extension of 3'-chromosomal termini with the 6-nucleotide telomeric repeat unit, 5'-TTAGGG-More Product Information