Anti-C9ORF103 Rabbit Polyclonal Antibody (ALEXA FLUOR® 350)
Supplier: Bioss
BS-15308R-A350
BOSSBS-15308R-A350EA 893 EUR
BOSSBS-15308R-A350
Anti-C9ORF103 Rabbit Polyclonal Antibody (ALEXA FLUOR® 350)
Antibodies
C9orf103 (chromosome 9 open reading frame 103), also known as gluconate kinase, is a 187 amino acid protein that belongs to the gluconokinase gntK/gntV family and catalyses the conversion of ATP and D-gluconate to ADP and 6-D-gluconate. Existing as three alternatively spliced isoforms, the gene encoding C9orf103 maps to human chromosome 9q21.32. Chromosome 9 consists of about 145 million bases, represents 4% of the human genome and encodes nearly 900 genes. Thought to play a role in gender determination, deletion of the distal portion of 9p can lead to development of male to female sexMore Product Information