Anti-HBsAg Goat Polyclonal Antibody (Cy7®)

Supplier: Bioss
BS-1557G-CY7
BOSSBS-1557G-CY7EA 883 EUR
BOSSBS-1557G-CY7
Anti-HBsAg Goat Polyclonal Antibody (Cy7®)
Antibodies
Hepatitis B Virus (HBV) infection induces a disease state characterised by liver damage, inflammation and viral persistence. Infection also increases the risk of hepatocellular carcinoma. HBV belongs to the Hepadnaviridae family of viruses. Its genome consists of partially double stranded circular DNA. The DNA is enclosed in a nucleocapsid, or core antigen (HBcAg), which is surrounded by a spherical envelope (surface antigen or HBsAg). The core antigen shares its sequences with the e antigen (HBeAg) but no cross reactivity between the two proteins has been observed. The HBV genome alsoMore Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable
Antigen name HBsAg
Antigen synonyms HBV major surface antigen|HBV surface antigen|HBsAg|Hepatitis B Virus major surface antigen|Major surface antigen|S|Human Hepatitis B Surface Antigen
Antibody type Primary
Clonality Polyclonal
Conjugation Cy7®
Reactivity Virus,Human,Mouse
Host Goat
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Modification Unmodified
Immunogen Human Hepatitis B Surface Antigen
Form Liquid
Molecular weight 44 kDa
Purification Purified by Protein A
Concentration 1 µg/µl
Storage buffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Storage temperature Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Shipping temperature 4 °C
Tested applications ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR