Anti-K19/CYFRA21-1 Rabbit Polyclonal Antibody (ALEXA FLUOR® 488)

Supplier: Bioss
BS-15590R-A488
BOSSBS-15590R-A488EA 876 EUR
BOSSBS-15590R-A488
Anti-K19/CYFRA21-1 Rabbit Polyclonal Antibody (ALEXA FLUOR® 488)
Antibodies
Involved in the organisation of myofibers. Together with KRT8, helps to link the contractile apparatus to dystrophin at the costameres of striated muscle.

Type: Primary
Antigen: K19/CYFRA21-1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 488
Public Immunogen Range: 301-400/400
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR