Anti-SKI Rabbit Polyclonal Antibody (Cy5.5®)
Supplier: Bioss
BS-1684R-CY5.5
BOSSBS-1684R-CY5.5EA 900 EUR
BOSSBS-1684R-CY5.5
Anti-SKI Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
SKI is a nuclear protein that forms homodimers and heterodimers and binds to DNA to function as transcriptional activators and repressors. The SKI oncoprotein dramatically affects cell growth, differentiation, and/or survival. SKI was shown to act in distinct signaling pathways including those involving nuclear receptors, transforming growth factor beta, and tumor suppressors. SKI inhibits transforming growth factor-beta (TGF-beta) signaling through interaction with Smad proteins. SKI represses Smad-mediated transcriptional activation, probably through its action as a transcriptional co-More Product Information