Anti-DCX Rabbit Polyclonal Antibody

Supplier: Bioss
BS-3113R
BOSSBS-3113REA 743 EUR
BOSSBS-3113R
Anti-DCX Rabbit Polyclonal Antibody
Antibodies
Neuronal Marker Doublecortin (DCX) is a microtubule-associated protein expressed almost exclusively in immature neurons. Neuronal precursors begin to express DCX shortly after exiting the cell cycle, and continue to express DCX for 2-3 weeks as the cells mature into neurons. Downregulation of DCX begins after 2 weeks, and occurs at the same time that these cells begin to express, a marker for mature neurons. Due to the nearly exclusive expression of DCX in developing neurons, this protein has been used increasingly as a marker for neurogenesis. Indeed, the levels of DCX expression increaseMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol DCX
Antigen name Doublecortin
Environmentally Preferable
Antigen synonyms LISX|Lis-X|DBCN|Lissencephalin-X|Doublin|Neuronal migration protein doublecortin|DCX
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 1641
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
Modification Ser128
Form liquid
Purification Purified by Protein A.
Concentration 1 ug/ul
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
Shipping temperature 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR