Anti-MAGE-A4 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Supplier: Bioss
BS-3645R-A680
BOSSBS-3645R-A680EA 890 EUR
BOSSBS-3645R-A680
Anti-MAGE-A4 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
This gene is a member of the MAGEA gene family. The members of this family encode proteins with 50 to 80% sequence identity to each other. The promoters and first exons of the MAGEA genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. The MAGEA genes are clustered at chromosomal location Xq28. They have been implicated in some hereditary disorders, such as dyskeratosis congenita. At least four variants encoding the same protein have been found for this gene.

Type: Primary
More Product Information
Order Now

SPECIFICATIONS

Antigen name MAGE-A4
Environmentally Preferable
Antigen synonyms MAGE X2 antigen|MAGE4A|Cancer/testis antigen 1.4|Melanoma associated antigen 4|MAGE 41 antigen|CT1.4|MAGE X2|Melanoma antigen family A 4|MAGE4B|MAGEA 4|MAGE 4A|MAGE 41|MAGEA4|MAGE 4B|MAGE41|MAGE 4|MAGE 4 antigen|MAGE4
Antibody type Primary
Clonality Polyclonal
Conjugation Alexa Fluor® 680
Reactivity
Host Rabbit
Gene ID 4103
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Modification Unmodified
Epitope 110-210/317
Immunogen KLH conjugated synthetic peptide derived from human MAGE-A4
Form Liquid
Molecular weight 35 kDa
Purification Purified by Protein A
Concentration 1 µg/µl
Storage buffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Storage temperature Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Shipping temperature 4 °C
Tested applications ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR