Anti-CDK11B Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss
BS-3999R-CY3
BOSSBS-3999R-CY3EA 900 EUR
BOSSBS-3999R-CY3
Anti-CDK11B Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Summary: This gene encodes a member of the p34Cdc2 protein kinase family. p34Cdc2 kinase family members are known to be essential for eukaryotic cell cycle control. This gene is in close proximity to CDC2L2, a nearly identical gene in the same chromosomal region. The gene loci including this gene, CDC2L2, as well as metalloprotease MMP21/22, consist of two identical, tandemly linked genomic regions which are thought to be a part of the larger region that has been duplicated. This gene and CDC2L2 were shown to be deleted or altered frequently in neuroblastoma with amplified MYCN genes. TheMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol CDK11B
Antigen name Cyclin-dependent Kinase 11B
Environmentally Preferable
Antigen synonyms p58CDC2L1|CDK11-p46|p58CLK-1|CLK-1|p58|PK58|CDK11|CDC2L1|CDK11-p58|PITSLREA|CDK11-p110|CD2L1
Antibody type Primary
Clonality Polyclonal
Conjugation Cy3®
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 984
Isotype IgG
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR