Anti-testis antigen 1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Supplier: Bioss
BS-4501R-A680
BOSSBS-4501R-A680EA 890 EUR
BOSSBS-4501R-A680
Anti-testis antigen 1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
The protein encoded by this gene is an antigen that is overexpressed in many cancers but that is also expressed in normal testis. This gene is found in a duplicated region of the X-chromosome and therefore has a neighboring gene of identical sequence.

Type: Primary
Antigen: testis antigen 1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 75-180/180
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen name testis antigen 1
Environmentally Preferable
Antigen synonyms Autoimmunogenic cancer/testis antigen NY-ESO-1|LAGE2B|CT6.1|LAGE2A|NY-ESO-1|LAGE2|CTAG1|L antigen family member 2|CTAG|ESO1|LAGE-2|Cancer/testis antigen 6.1|CTAG1A|Cancer/testis antigen 1
Antibody type Primary
Clonality Polyclonal
Conjugation Alexa Fluor® 680
Reactivity Human
Host Rabbit
Gene ID 1485
Isotype IgG
ImmunoChemistry Yes
ImmunoFluorescence Yes
Modification Unmodified
Epitope 75-180/180
Immunogen KLH conjugated synthetic peptide derived from human Cancer/testis antigen 1B/1A
Form Liquid
Molecular weight 18 kDa
Purification Purified by Protein A
Concentration 1 µg/µl
Storage buffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Storage temperature Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Shipping temperature 4 °C
Tested applications ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR