Anti-ENPP1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss
BS-4913R-A488
BOSSBS-4913R-A488EA 893 EUR
BOSSBS-4913R-A488
Anti-ENPP1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
By generating PPi, plays a role in regulating pyrophosphate levels, and functions in bone mineralization and soft tissue calcification. PPi inhibits mineralization by binding to nascent hydroxyapatite (HA) crystals, thereby preventing further growth of these crystals. Preferentially hydrolyzes ATP, but can also hydrolyze other nucleoside 5' triphosphates such as GTP, CTP, TTP and UTP to their corresponding monophosphates with release of pyrophosphate and diadenosine polyphosphates, and also 3',5'-cAMP to AMP. May also be involved in the regulation of the availability of nucleotide sugars inMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol ENPP1
Antigen name Ectonucleotide pyrophosphatase/phosphodiesterase 1
Environmentally Preferable
Antigen synonyms PCA1|phosphodiesterase I/nucleotide pyrophosphatase 1|M6S1|membrane component|surface marker 1|plasma-cell membrane glycoprotein 1|alkaline phosphodiesterase 1|ENPP1|HGNC:3356|PDNP1|Ly-41 antigen|PC-1|NPP1|NPPS|ectonucleotide pyrophosphatase/phosphodiesterase 1|OTTHUMP00000043194|chromosome 6
Antibody type Primary
Clonality Polyclonal
Conjugation Alexa Fluor® 488
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 5167
Isotype IgG
ImmunoChemistry Yes
Immunogen 97-150/925
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR