Anti-JUN Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss
BS-5458R-CY3
BOSSBS-5458R-CY3EA 930 EUR
BOSSBS-5458R-CY3
Anti-JUN Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Transcription factor that recognizes and binds to the enhancer heptamer motif 5'-TGA[CG]TCA-3'. Promotes activity of NR5A1 when phosphorylated by HIPK3 leading to increased steroidogenic gene expression upon cAMP signaling pathway stimulation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: JUN
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR