Anti-NFKB p65 Ser529 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Supplier: Bioss
BS-5660R-A680
BOSSBS-5660R-A680EA 921 EUR
BOSSBS-5660R-A680
Anti-NFKB p65 Ser529 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
NF-kappa-B is a ubiquitous transcription factor involved in several biological processes. It is held in the cytoplasm in an inactive state by specific inhibitors. Upon degradation of the inhibitor, NF-kappa-B moves to the nucleus and activates transcription of specific genes. NF-kappa-B is composed of NFKB1 or NFKB2 bound to either REL, RELA, or RELB. The most abundant form of NF-kappa-B is NFKB1 complexed with the product of this gene, RELA. Four transcript variants encoding different isoforms have been found for this gene.

Type: Primary
Antigen: NFKB p65 Ser529
Clonality: Polyclonal
Clone:More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR