Anti-FTH1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss
BS-5907R-A647
BOSSBS-5907R-A647EA 915 EUR
BOSSBS-5907R-A647
Anti-FTH1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
Stores iron in a soluble, non-toxic, readily available form. Important for iron homeostasis. Has ferroxidase activity. Iron is taken up in the ferrous form and deposited as ferric hydroxides after oxidation. Also plays a role in delivery of iron to cells. Mediates iron uptake in capsule cells of the developing kidney (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Ferritin heavy chain
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR