Anti-BAZ1B Rabbit Polyclonal Antibody

Supplier: Bioss
BS-6128R
BOSSBS-6128REA 725 EUR
BOSSBS-6128R
Anti-BAZ1B Rabbit Polyclonal Antibody
Antibodies
This gene encodes a member of the bromodomain protein family. The bromodomain is a structural motif characteristic of proteins involved in chromatin-dependent regulation of transcription. This gene is deleted in Williams-Beuren syndrome, adevelopmental disorder caused by deletion of multiple genes at 7q11.23.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: BAZ1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR