Anti-NEK3 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Supplier: Bioss
BS-6261R-A555
BOSSBS-6261R-A555EA 893 EUR
BOSSBS-6261R-A555
Anti-NEK3 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
This gene encodes a member of the NimA (never in mitosis A) family of serine/threonine protein kinases. The encoded protein differs from other NimA family members in that it is not cell cycle regulated and is found primarily in the cytoplasm. The kinase is activated by prolactin stimulation, leading to phosphorylation of VAV2 guanine nucleotide exchange factor, paxillin, and activation of the RAC1 GTPase. Multiple transcript variants encoding different isoforms have been found for this gene.[provided by RefSeq, Mar2009].
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen:More Product Information