Anti-SLC2A10 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss
BS-6325R-CY5
BOSSBS-6325R-CY5EA 900 EUR
BOSSBS-6325R-CY5
Anti-SLC2A10 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Defects in SLC2A10 are the cause of arterial tortuosity syndrome (ATS) [MIM:208050]. ATS is an autosomal recessive disorder characterized by tortuosity and elongation of major arteries, often resulting in death at young age. Other typical features include aneurysms of large arteries and stenosis of the pulmonary artery, in association with facial features and several connective tissue manifestations such as soft skin and joint laxity. Histopathological findings include fragmentation of elastic fibers in the tunica media of large arteries.

Recommended Dilutions: IF(IHC-P): 1:50-200

Order Now

SPECIFICATIONS

Antigen symbol SLC2A10
Environmentally Preferable
Antigen name Solute Carrier Family 2 Member A10
Antigen synonyms GLUT10|ATS
Antibody type Primary
Clonality Polyclonal
Conjugation Cy5®
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 81031
Isotype IgG
ImmunoChemistry Yes
Immunogen 299-316/541
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR