Anti-Sortilin Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Supplier: Bioss
BS-6329R-A680
BOSSBS-6329R-A680EA 877 EUR
BOSSBS-6329R-A680
Anti-Sortilin Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
Functions as a sorting receptor in the Golgi compartment and as a clearance receptor on the cell surface. Required for protein transport from the Golgi apparatus to the lysosomes by a pathway that is independent of the mannose-6-phosphate receptor (M6PR). Also required for protein transport from the Golgi apparatus to the endosomes. Promotes neuronal apoptosis by mediating endocytosis of the proapoptotic precursor forms of BDNF (proBDNF) and NGFB (proNGFB). Also acts as a receptor for neurotensin. May promote mineralisation of the extracellular matrix during osteogenic differentiation byMore Product Information